Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diisobutyl hydrogen phosphate | 6303-30-6 | MFCD25965560 | 98.0% | 210.21 g/mol | C8H19O4P | 5 MG
SDP

Catalog No. 5000284563
Encompass_Preferred

Diisobutyl hydrogen phosphate is a dialkyl phosphate ester used primarily as a flame retardant and for research applications. It is a low-molecular-weight organophosphate supplied in research-scale quantities and typically offered at high purity for analytical and assay use.

  • Dialkyl phosphate ester with formula C8H19O4P.
  • Functions as a flame retardant and biochemical reagent.
  • Molecular weight approximately 210.21 g/mol.
  • Available in small quantities suitable for analytical work.
  • High purity appropriate for assay and characterization.
  • CAS number 6303-30-6 for unambiguous identification.

Catalog No. 50-002-84563 Supplier Medchemexpress LLC Supplier No. HYW5858845MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diisobutyl hydrogen phosphate is a dialkyl phosphate ester used primarily as a flame retardant and for research applications. It is a low-molecular-weight organophosphate supplied in research-scale quantities and typically offered at high purity for analytical and assay use.

  • Dialkyl phosphate ester with formula C8H19O4P.
  • Functions as a flame retardant and biochemical reagent.
  • Molecular weight approximately 210.21 g/mol.
  • Available in small quantities suitable for analytical work.
  • High purity appropriate for assay and characterization.
  • CAS number 6303-30-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.