Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DiIsobutyl Ketone (DIBK), Reagents

Catalog No. 01337279
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-279 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-279 Supplier Reagents Holdings Llc Supplier No. C219645020E6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DiIsobutyl Ketone (DIBK), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 108-83-8
Boiling Range 166°C
Melting Point -41.5°C
Color Colorless
Boiling Point 166°C
Melting Range -41.5°C
Packaging Steel Pail
Molecular Formula C9H18O
Quantity 20 L
Specific Gravity 0.808
Synonym 2,6-Dimethyl-4-heptanone
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
SMILES CC(C)CC(=O)CC(C)C
IUPAC Name 2,6-dimethylheptan-4-one
Molecular Weight (g/mol) 142.24
Grade Reagent
Physical Form Liquid
Chemical Name or Material DiIsobutyl Ketone (DIBK)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.