Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminium chloride, 0.8M solution in heptane, AcroSeal™

Catalog No. AC377561000
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 1779-25-5 | C8H18AlCl | 176.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-25-5, 142-82-5
Molecular Formula C8H18AlCl
Molecular Weight (g/mol) 176.67
MDL Number MFCD00008927
InChI Key HQMRIBYCTLBDAK-UHFFFAOYSA-M
Synonym diisobutylaluminum chloride, chlorodiisobutylaluminium, chlorodiisobutylaluminum, diisobutylchloroaluminum, aluminum, chlorobis 2-methylpropyl, aluminum, chlorodiisobutyl, bis isobutyl aluminum chloride, diisobutylaluminum monochloride, chlorobis 2-methylpropyl aluminum, alluminio diisobutil-monocloruro italian
PubChem CID 15688
IUPAC Name chloro-bis(2-methylpropyl)alumane
SMILES CC(C)C[Al](CC(C)C)Cl

Specifications

Density 0.7260g/mL
Flash Point −4°C
Packaging AcroSeal™ Glass bottle
Quantity 100 mL
Specific Gravity 0.726
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 176.67
Chemical Name or Material Diisobutylaluminium chloride
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases
missing translation for 'healthHazard3' GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Im
EINECSNumber 217-216-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.