Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™

Catalog No. p-3586422
Change view
Click to view available options
Quantity:
100 mL
400 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201081000 100 mL AcroSeal™ Glass bottle
AC201084000 400 mL AcroSeal™ Glass bottle
AC201088000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC201081000 Supplier Thermo Scientific Chemicals Supplier No. 201081000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1191-15-7 | C8H19Al | 142.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisobutylaluminium hydride
CAS 1191-15-7, 108-88-3
Density 0.8480g/mL
Boiling Point 110.0°C
Flash Point 4°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H19Al
Linear Formula [(CH3)2CHCH2]2AIH
Quantity 100 mL
MDL Number MFCD00008928
Beilstein 04, IV, 4400
Fieser 01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
Specific Gravity 0.848
Merck Index 15, 3212
Synonym DIBAL-H
Solubility Information Solubility in water: reacts violently. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Molecular Weight (g/mol) 142.22
Formula Weight 142.22
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.