Learn More
Diisobutylaluminum hydride, 25% w/w in hexane, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Colorless |
| Density | 0.701 g/mL |
| Linear Formula | [(CH3)2CHCH2]2AlH |
| Quantity | 250 mL |
| UN Number | UN3399 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 142.22 |
| Physical Form | Liquid |
| Chemical Name or Material | Diisobutylaluminum hydride |
Safety and Handling
| Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361f-H373 |
| Hazard Statement | GHS H Statement H224-H260-H304-H361-H373-H314-H318-H336 Extremely flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure. Causes severe skin burns and eye damage. Causes serious eye damage. May cause drowsiness or dizziness. |
| Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.