Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminum hydride, 25% w/w in hexane, Thermo Scientific™

Catalog No. AA42588K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42588K2 1 L
AA42588AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42588K2 Supplier Thermo Scientific Chemicals Supplier No. 42588K2

missing translation for 'validMainframeMsg'

Reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928
Synonym DIBAL-H

Specifications

Color Colorless
Density 0.701 g/mL
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 1 L
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H224-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.