Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisobutylaluminum Hydride

Catalog No. 2568111KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-681-11KG 1 kg
25-681-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-681-11KG Supplier Sigma Aldrich Supplier No. 2568111KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisobutylaluminum hydride is commonly used as a reducing agent in organic synthesis.Diisobutylaluminum hydride can be used in the following protocols:As a promotor of Tishchenko reaction of aldehydes. Conversion of benzylidene acetals of 1,2-and 1,3-glycols to the corresponding monobenzyl ethers of the glycols. To generate a novel chiral reducing agent based on (S)-proline for the asymmetric reduction of prochiral ketones.

Specifications

CAS 1191-15-7
Density 0.798 g/mL at 25°C
Boiling Point 116°C to 118°C (1 mmHg, lit.)
Molecular Formula C8H19Al
Linear Formula [(CH3)2CHCH2]2AlH
MDL Number MFCD00008928
Quantity 1 kg
Synonym DIBAL; DIBAL-H
Molecular Weight (g/mol) 142.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.