Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisobutylaluminum Hydride Solution

Catalog No. 192724700G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
4 x 25 g
700 g
15 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-272-4700G 700 g
19-272-4100G 100 g
19-272-44X25G 4 x 25 g
19-272-415KG 15 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 19-272-4700G Supplier Sigma Aldrich Supplier No. 192724700G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisobutylaluminum hydride reducing agent is commonly used for the reduction of an ester to aldehyde.Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1191-15-7
Density 0.846 g/mL at 25°C
Molecular Formula C8H19Al
Linear Formula [(CH3)2CHCH2]2AlH
MDL Number MFCD00008928
Quantity 700 g
Synonym DIBAL; DIBAL-H
Molecular Weight (g/mol) 142.22 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.