Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisobutylene

Catalog No. 381801LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-180-1LF 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 38-180-1LF Supplier Sigma Aldrich Supplier No. 381801LF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisobutylene is an acyclic alkene. It is composed of a mixture of 2,4,4-trimethyl-1-pentene and 2,4,4- trimethyl-2-pentene (isomeric forms of diisobutylene). It is isostructural to to iso-octane. It can be synthesized from tertiary butyl alcohol.Diisobutylene, an alkene, may be used in the following studies:To compose alternalte diesel fuel. As fuel in single-cylinder engine; exhaust generated from combustion due to the burning of fuel has been reported to contain high concentrations of olefins.1 Preparation of mono- and dialkylated diphenylamines.

Specifications

CAS 60835-71-4
Melting Point 76°C to 78°C
Molecular Formula C14H21NO5
Refractive Index n20/D 1.411
Linear Formula C14H21NO5
MDL Number MFCD00068686
Quantity 500 mg
Synonym 2,4,4-Trimethyl-1-pentene + 2,4,4-Trimethyl-2-pentene
Molecular Weight (g/mol) 283.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.