Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisoctyl Adipate, MP Biomedicals

Catalog No. ICN208052 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208052 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208052 Supplier MP Biomedicals, Inc Supplier No. 0520805280

Chemical Identifiers

CAS 1330-86-5
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.574
InChI Key CJFLBOQMPJCWLR-UHFFFAOYSA-N
Synonym diisooctyl adipate, bis 6-methylheptyl adipate, hexanedioic acid, diisooctyl ester, diiosoctyl adipate, unii-1pn0d86ixj, diisooctyladipate, plasthall dioa, monoplex dioa, diisoctyl adipate, dsstox_cid_7388
PubChem CID 66932
IUPAC Name bis(6-methylheptyl) hexanedioate
SMILES CC(C)CCCCCOC(=O)CCCCC(=O)OCCCCCC(C)C

Specifications

Quantity 100 g
Chemical Name or Material Diisoctyl Adipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.