Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisodecyl 4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™
SDP

Catalog No. T060325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060325ML 25 mL
T0603500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T060325ML Supplier TCI America Supplier No. T060325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87826-26-4
Molecular Formula C28H50O4
Molecular Weight (g/mol) 450.704
MDL Number MFCD00070615
InChI Key XRFYGUOAWVKOCQ-UHFFFAOYSA-N
Synonym 4-Cyclohexene-1,2-dicarboxylic Acid Diisodecyl Ester, Diisodecyl 1,2,3,6-Tetrahydrophthalate, 1,2,3,6-Tetrahydrophthalic Acid Diisodecyl Ester
PubChem CID 587973
IUPAC Name bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate
SMILES CC(C)CCCCCCCOC(=O)C1CC=CCC1C(=O)OCCCCCCCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 450.70
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Diisodecyl 4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.