Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisodecyl Adipate, TCI America™
SDP

Catalog No. A016425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016425ML 25 mL
A0164500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016425ML Supplier TCI America Supplier No. A016425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27178-16-1
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.682
MDL Number MFCD00048392
InChI Key YKGYQYOQRGPFTO-UHFFFAOYSA-N
Synonym diisodecyl adipate, hexanedioic acid, diisodecyl ester, adipic acid diisodecyl ester, monoplex dda, adipic acid, diisodecyl ester, bis 8-methylnonyl hexanedioate, diisodecyl adipate, adipate, diisodecyl hexanedioate, hexanedioic acid, 1,6-diisodecyl ester, dsstox_cid_7924
PubChem CID 33733
IUPAC Name bis(8-methylnonyl) hexanedioate
SMILES CC(C)CCCCCCCOC(=O)CCCCC(=O)OCCCCCCCC(C)C

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 mL
Formula Weight 426.68
Physical Form Liquid
Chemical Name or Material Diisodecyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV1485000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.