Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisononyl Cyclohexanedicarboxylate (mixture of isomers), TCI America™
SDP

Catalog No. D375425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D375425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D375425ML Supplier TCI America Supplier No. D375425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 508181-38-2
Molecular Formula C78H144O12
Molecular Weight (g/mol) 1274.00
MDL Number MFCD28100825
InChI Key QSNOUBLJFWGBSD-UHFFFAOYNA-N
Synonym Cyclohexanedicarboxylic Acid Diisononyl Ester
PubChem CID 131674806
IUPAC Name 1,2-bis(3,5,5-trimethylhexyl) cyclohexane-1,2-dicarboxylate; 1,3-bis(3,5,5-trimethylhexyl) cyclohexane-1,3-dicarboxylate; 1,4-bis(3,5,5-trimethylhexyl) cyclohexane-1,4-dicarboxylate
SMILES CC(CCOC(=O)C1CCC(CC1)C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C.CC(CCOC(=O)C1CCCC(C1)C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C.CC(CCOC(=O)C1CCCCC1C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 424.67
Physical Form Liquid
Chemical Name or Material Diisononyl Cyclohexanedicarboxylate (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.