Learn More
Medchemexpress LLC Diisononyl phthalate 1kg | 28553-12-0 | 418.61 g/mol | C26H42O4 | 1 KG
SDP
Catalog No. 5000308242
Encompass_Preferred
Diisononyl phthalate (DINP) is a phthalate ester used as a plasticizer to increase flexibility in polymers and for research and analytical applications. It is commonly used in resin solvents, building materials, lubricants, adhesives, cosmetics, and printing inks.
  • Suitable for research and analytical use.
  • Molecular formula C26H42O4 and molecular weight 418.61 g/mol for identification.
  • Soluble at ≥ 5 mg/mL in common solvents, facilitating stock solution preparation.
  • Commonly used as a plasticizer and solvent in material formulation.
  • Available in bulk quantities for formulation and testing.
Catalog No. 50-003-08242 Supplier Medchemexpress LLC Supplier No. HY1164391KG
Diisononyl phthalate (DINP) is a phthalate ester used as a plasticizer to increase flexibility in polymers and for research and analytical applications. It is commonly used in resin solvents, building materials, lubricants, adhesives, cosmetics, and printing inks.
  • Suitable for research and analytical use.
  • Molecular formula C26H42O4 and molecular weight 418.61 g/mol for identification.
  • Soluble at ≥ 5 mg/mL in common solvents, facilitating stock solution preparation.
  • Commonly used as a plasticizer and solvent in material formulation.
  • Available in bulk quantities for formulation and testing.