Learn More
Medchemexpress LLC Diisononyl phthalate 25g | 28553-12-0 | 25 G
SDP
Catalog No. 5000227107
Encompass_Preferred
Diisononyl phthalate (DINP) is a phthalate ester used as a plasticizer to increase flexibility and durability of polymer formulations. It is provided in research-sized packages for formulation testing and comes with supporting documentation (SDS and datasheet).
  • Plasticizer for polymers, resins, and PVC formulations.
  • Improves flexibility, impact resistance, and durability.
  • Compatible with common organic solvents and resin systems.
  • Available in small research quantities for formulation and testing.
  • Delivered with CAS identification and safety documentation (SDS).
Catalog No. 50-002-27107 Supplier Medchemexpress LLC Supplier No. HY11643925G
Diisononyl phthalate (DINP) is a phthalate ester used as a plasticizer to increase flexibility and durability of polymer formulations. It is provided in research-sized packages for formulation testing and comes with supporting documentation (SDS and datasheet).
  • Plasticizer for polymers, resins, and PVC formulations.
  • Improves flexibility, impact resistance, and durability.
  • Compatible with common organic solvents and resin systems.
  • Available in small research quantities for formulation and testing.
  • Delivered with CAS identification and safety documentation (SDS).