Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisononyl phthalate

Catalog No. 3766631L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-666-31L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-666-31L Supplier Sigma Aldrich Supplier No. 3766631L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisononyl phthalate(DINP) is a plasticizer with a combination of 9-carbon branched dialkyl phthalate isomers. It can plasticize a variety of polyvinylchloride (PVC) based products that are majorly used in personal care items, nutritional supplements, and pharmaceutical industries.

Specifications

CAS 28553-12-0
Density 0.972 g/mL (at 25°C)
Molecular Formula C26H42O4
Linear Formula C6H4(CO2C9H19)2
Refractive Index n20/D 1.485 (literature)
Quantity 1 L
Synonym DINP
Molecular Weight (g/mol) 418.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.