Learn More
Medchemexpress LLC Diisononyl phthalate 500g | 28553-12-0 | 418.6 g/mol | C26H42O4 | 500 G
SDP
Catalog No. 5000226179
Encompass_Preferred
Diisononyl phthalate is a phthalate ester used as a plasticizer for polymers and coatings. It is an oily, colorless liquid, denser than water and insoluble in water. Common applications include PVC formulations, resin solvents, adhesives, lubricants, cosmetics, and printing inks.
  • Common plasticizer for PVC and other polymers
  • Oily, colorless liquid; low water solubility
  • Used in adhesives, lubricants, and printing inks
  • Applicable for resin solvents and building materials
  • CAS 28553-12-0; molecular formula C26H42O4
Catalog No. 50-002-26179 Supplier Medchemexpress LLC Supplier No. HY116439500G
Diisononyl phthalate is a phthalate ester used as a plasticizer for polymers and coatings. It is an oily, colorless liquid, denser than water and insoluble in water. Common applications include PVC formulations, resin solvents, adhesives, lubricants, cosmetics, and printing inks.
  • Common plasticizer for PVC and other polymers
  • Oily, colorless liquid; low water solubility
  • Used in adhesives, lubricants, and printing inks
  • Applicable for resin solvents and building materials
  • CAS 28553-12-0; molecular formula C26H42O4