Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisononyl Phthalate (mixture of branched chain isomers), TCI America™
SDP

Catalog No. P0300500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0300500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0300500G Supplier TCI America Supplier No. P0300500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28553-12-0
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.618
InChI Key HBGGXOJOCNVPFY-UHFFFAOYSA-N
Synonym Phthalic Acid Diisononyl Ester, DINP
PubChem CID 590836
ChEBI CHEBI:35459
IUPAC Name bis(7-methyloctyl) benzene-1,2-dicarboxylate
SMILES CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC(C)C

Specifications

Physical Form Liquid
Color Colorless
Melting Point -43°C
Boiling Point 252°C
Quantity 500 g
Formula Weight 418.62
Chemical Name or Material Diisononyl Phthalate (mixture of branched chain isomers)
EINECSNumber (3)-1307
RTECSNumber CZ3395000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.