Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopentyl phthalate, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018586 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8586 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8586 Supplier MilliporeSigma Supelco Supplier No. 44263250MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 605-50-5
Linear Formula C18H26O4
MDL Number MFCD00127736
Synonym 1,2-Benzenedicarboxylic acid 1,2-bis(3-methylbutyl) ester; Bis(3-methylbutyl) phthalate; Diisoamyl phthalate; NSC 17070; Phthalic acid diisopentyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 306.40
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.