Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisopropyl Azodicarboxylate

Catalog No. 225541100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-1100G 100 g
22-554-15G 5 g
22-554-1500G 500 g
22-554-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 22-554-1100G Supplier Sigma Aldrich Supplier No. 225541100G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reacted with dioxaphosphorinanes and iron catalyst to generate rearranged phosphonium ylide products.Reactant for preparation of:Chromenes resembling classical cannabinoidsMK-3281 inhibitor of the hepatitis C virus NS5B polymeraseNorbornene-based guanidine-rich polymers as mimcs of cell-penetrating peptides (CPP)Analogues of the Pseudomonas aeruginosa quorum-sensing molecule N-(3-Oxododecanoyl)-l-homoserine lactone with immunosuppressive but non-LasR-inducing propertiesAcceptor-donor-acceptor organic dyes for dye-sensitized solar cells1,3-dioxane-2-carboxylic acid derivatives as PPAR+a/+a dual agonistsHydrazinophosphonates and hydrazinobisphosphonates via Mitsunobu and Arbuzov-type multicomponent application of the Morrison-Brunn-Huisgen betaine

Specifications

CAS 2446-83-5
Density 1.027 g/mL at 25°C
Boiling Point 75°C (0.25 mmHg, lit.)
Molecular Formula C8H14N2O4
Refractive Index n20/D 1.420
Linear Formula (CH3)2CHOOCN=NCOOCH(CH3)2
Quantity 100 g
MDL Number MFCD00008875
Synonym DIAD; Diisopropyl azodiformate
Molecular Weight (g/mol) 202.21 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.