Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl ether, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020398
Click to view available options
Quantity:
1 mL
5 mL
10 mL

Diisopropyl ether is a secondary ether , which finds applications as an extractant and oxygenate gasoline additive.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 827°F
CAS 108-20-3
Vapor Pressure 120 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.725 g/mL (at 25°C)
Flash Point -29°C
Physical Form Neat
Linear Formula (CH3)2 CHOCH(CH3)2
MDL Number MFCD00008880
Melting Point -85°C
Boiling Point 68°C to 69°C
Synonym Isopropyl ether
Type Analytical Standard
Quantity 1 mL
Formula Weight 102.17
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.