Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Ether Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002417 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2417 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2417 Supplier MilliporeSigma Supelco Supplier No. PHR21833X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 108-20-3
Ignition Point 827°F
Vapor Pressure 120 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.725 g/mL (at 25°C)
Flash Point -48°C
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula (CH3)2 CHOCH(CH3)2
MDL Number MFCD00008880
Melting Point -85°C
Boiling Point 68°C to 69°C
Synonym Diisopropyl ether; Isopropyl ether
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 102.17
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.