Learn More
Sigma Aldrich Fine Chemicals Biosciences Diisopropyl ether puriss.
Catalog No. 502899391
Encompass
Diisopropyl ether is a colorless liquid. It is widely used as a solvent for the isolation of fats and oils. It can be synthesized from isopropyl alcohol. Its mixture with isooctane has been added to the gasoline fuel as an antiknocking agent. Its synthesis from propylene and water in a single-pass trickle-bed reactor in the presence of an ion-exchange resin catalyst Amberlyst 15 has been proposed.
Catalog No. 50-289-9391 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 382702.5LF
Diisopropyl ether is a colorless liquid. It is widely used as a solvent for the isolation of fats and oils. It can be synthesized from isopropyl alcohol. Its mixture with isooctane has been added to the gasoline fuel as an antiknocking agent. Its synthesis from propylene and water in a single-pass trickle-bed reactor in the presence of an ion-exchange resin catalyst Amberlyst 15 has been proposed.