Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl methylphosphonate, 90%, Thermo Scientific Chemicals

Catalog No. AC449381000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449381000 100 g
AC449380050 5 g
AC449380250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449381000 Supplier Thermo Scientific Chemicals Supplier No. 449381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-75-6
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.18
MDL Number MFCD00015022
InChI Key WOAFDHWYKSOANX-UHFFFAOYSA-N
Synonym diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
PubChem CID 3073
ChEBI CHEBI:77325
IUPAC Name 2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(C)(=O)OC(C)C

Specifications

Density 0.9850g/mL
Boiling Point 50.0°C to 51.0°C (1.0 mmHg)
Assay Percent Range 88% min. (GC)
Packaging Glass Bottle
Linear Formula [(CH3)2C(H)O]2P(O)CH3
Quantity 100 g
Specific Gravity 0.985
Formula Weight 180.18
Percent Purity 90%
Chemical Name or Material Diisopropyl Methylphosphonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 215-896-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.