Learn More
Medchemexpress LLC Diisopropyl phthalate | 605-45-8 | 99.4% | 250.29 | 10 MG
SDP
Catalog No. 5000452598
Encompass_Preferred
Diisopropyl phthalate (DiPP) is a phthalate diester, commonly found as a colorless to light yellow liquid with a density of 1.063 g/cm3. It is widely used as an additive in plastics and consumer products. Diisopropyl phthalate has been detected in honey and royal jelly and exhibits a weak binding ability to the recombinant human estrogen receptor.
  • Widely used as an additive in plastics and consumer products
  • Detected in honey and royal jelly
  • Exhibits weak binding to the recombinant human estrogen receptor
  • Available in various quantities including liquid and solution forms
Catalog No. 50-004-52598 Supplier Medchemexpress LLC Supplier No. HYW011926S10MG
Diisopropyl phthalate (DiPP) is a phthalate diester, commonly found as a colorless to light yellow liquid with a density of 1.063 g/cm3. It is widely used as an additive in plastics and consumer products. Diisopropyl phthalate has been detected in honey and royal jelly and exhibits a weak binding ability to the recombinant human estrogen receptor.
  • Widely used as an additive in plastics and consumer products
  • Detected in honey and royal jelly
  • Exhibits weak binding to the recombinant human estrogen receptor
  • Available in various quantities including liquid and solution forms