Learn More
Medchemexpress LLC Diisopropyl phthalate | 605-45-8 | 99.4% | 250.29 | 5 G
SDP
Catalog No. 5000452364
Encompass_Preferred
Diisopropyl phthalate (DiPP) is a phthalate diester, which is widely used as an additive in plastics and consumer products. It has a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM.
  • Widely used as an additive in plastics
  • Incorporated into consumer products
  • Exhibits weak binding ability to the recombinant human estrogen receptor
Catalog No. 50-004-52364 Supplier Medchemexpress LLC Supplier No. HYW0119265G
Diisopropyl phthalate (DiPP) is a phthalate diester, which is widely used as an additive in plastics and consumer products. It has a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM.
  • Widely used as an additive in plastics
  • Incorporated into consumer products
  • Exhibits weak binding ability to the recombinant human estrogen receptor