Learn More
Medchemexpress LLC Diisopropyl phthalate-d4 (DiPP-d4) | 2708282-44-2 | 98.8% | 254.31 | 5 MG
SDP
Catalog No. 5000452905
Encompass_Preferred
Diisopropyl phthalate-d4 (DiPP-d4) is the deuterium-labeled version of Diisopropyl Phthalate. Diisopropyl phthalate (DiPP) is a phthalate diester commonly used as an additive in plastics and consumer products. It has a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM. Deuterium-labeled compounds, such as Diisopropyl phthalate-d4, incorporate stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily for use as tracers in quantitative analysis during drug development.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52905 Supplier Medchemexpress LLC Supplier No. HYW011926S5MG
Diisopropyl phthalate-d4 (DiPP-d4) is the deuterium-labeled version of Diisopropyl Phthalate. Diisopropyl phthalate (DiPP) is a phthalate diester commonly used as an additive in plastics and consumer products. It has a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM. Deuterium-labeled compounds, such as Diisopropyl phthalate-d4, incorporate stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily for use as tracers in quantitative analysis during drug development.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.