Learn More
Medchemexpress LLC Diisopropyl phthalate (DiPP) | 605-45-8 | 99.4% | 250.29 | 1 ML
SDP
Catalog No. 5000452363
Encompass_Preferred
Diisopropyl phthalate (DiPP) is a phthalate diester, widely used as an additive in plastics and consumer products. It exhibits a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM.
  • Phthalate diester
  • Used as an additive in plastics and consumer products
  • Weak binding ability to the recombinant human estrogen receptor (IC50 of 41000 μM)
  • Appearance: Liquid (Density: 1.063 g/cm3)
  • Colorless to light yellow
Catalog No. 50-004-52363 Supplier Medchemexpress LLC Supplier No. HYW01192610MM/1ML
Diisopropyl phthalate (DiPP) is a phthalate diester, widely used as an additive in plastics and consumer products. It exhibits a weak binding ability to the recombinant human estrogen receptor, with an IC50 of 41000 μM.
  • Phthalate diester
  • Used as an additive in plastics and consumer products
  • Weak binding ability to the recombinant human estrogen receptor (IC50 of 41000 μM)
  • Appearance: Liquid (Density: 1.063 g/cm3)
  • Colorless to light yellow