Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Succinate 99.0+%, TCI America™
SDP

Catalog No. S0269500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0269500ML 500 mL
S026925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0269500ML Supplier TCI America Supplier No. S0269500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 924-88-9
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00059360
InChI Key YPLYFEUBZLLLIY-UHFFFAOYSA-N
Synonym Succinic Acid Diisopropyl Ester
PubChem CID 70213
IUPAC Name 1,4-bis(propan-2-yl) butanedioate
SMILES CC(C)OC(=O)CCC(=O)OC(C)C

Specifications

Color Colorless
Boiling Point 228°C
Quantity 500 mL
Formula Weight 202.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Succinate
EINECSNumber (2)-0848
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.