Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Sulfite 98.0+%, TCI America™
SDP

Catalog No. D297425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D297425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D297425G Supplier TCI America Supplier No. D297425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4773-13-1
Molecular Formula C6H14O3S
Molecular Weight (g/mol) 166.24
MDL Number MFCD06200838
InChI Key IIKLEMKAKOISSP-UHFFFAOYSA-N
PubChem CID 536816
IUPAC Name bis(propan-2-yl) sulfite
SMILES CC(C)OS(=O)OC(C)C

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 166.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Sulfite
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.