Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisopropylamine

Catalog No. 471224500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 x 100 mL
18 L
4 x 2.5 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-122-4500ML 500 mL
47-122-4 4 x 100 mL
47-122-425 4 x 2.5 L
47-122-425L 2.5 L
47-122-4100ML 100 mL
47-122-418L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 47-122-4500ML Supplier Sigma Aldrich Supplier No. 471224500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisopropylamine

Specifications

CAS 108-18-9
Melting Point -61°C
Density 0.722 g/mL at 25°C
Boiling Point 84°C
Molecular Formula C6H15N
Refractive Index n20/D 1.392
Linear Formula (CH3)2CHNHCH(CH3)2
MDL Number MFCD00008862
Quantity 500 mL
Synonym DIPA
Molecular Weight (g/mol) 101.19 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.