Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisopropylamine

Catalog No. 382901LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-290-1LF 1 L
38-290-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-290-1LF Supplier Sigma Aldrich Supplier No. 382901LF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisopropylamine

Specifications

CAS 108-18-9
Melting Point -61°C
Density 0.722 g/mL at 25°C
Boiling Point 84°C
Molecular Formula C6H15N
Refractive Index n20/D 1.392; n20/D 1.393
Linear Formula (CH3)2CHNHCH(CH3)2
MDL Number MFCD00008862
Quantity 1 L
Synonym DIPA
Molecular Weight (g/mol) 101.19 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.