Learn More
Hach Company Diisopropylamine, 99%, 108-18-9, MFCD00008862, 1L
Supplier:  Hach Company 2834453
Encompass
Linear Formula: (CH3)2CHNHCH(CH3)2, Molecular Weight: 101.19. Diisopropylamine (DIPA) reagent used to control pH when measuring Sodium with Hach 9245 and 9240 Sodium process analyzers. 1L can last up to 100 days.
Catalog No. NC1328893