Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pfaltz & Bauer DIISOPROPYLAMINE 99.5%

Catalog No. 507490087
Encompass

Diisopropylamine 99.5%; 500g; CAS: 108-18-9

Catalog No. 50-749-0087 Supplier Pfaltz & Bauer Supplier No. D32440500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diisopropylamine 99.5%; 500g; CAS: 108-18-9

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.