Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylamine, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019262 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9262 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9262 Supplier MilliporeSigma Supelco Supplier No. 382925MLF
Add to Cart
Add to Cart

Diisopropylamine, an aliphatic secondary amine, is a well-known precursor of lithium diisopropylamide (LDA), which is a hindered, strong, non-nucleophilic base utilized in various organic reactions.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 599°F
CAS 7664-93-9
Vapor Pressure 50 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.722 g/mL (at 25°C)
Flash Point -17°C - Closed Cup
Physical Form Neat
Linear Formula (CH3)2 CHNHCH(CH3)2
MDL Number MFCD00008862
Melting Point -61°C
Boiling Point 84°C
Synonym DIPA
Solubility Soluble in water
Type Analytical Standard
Quantity 5 mL
Formula Weight 101.19
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.