Learn More
Medchemexpress LLC Diisopropylammonium dichloroacetate | 660-27-5 | 98.0% | 500 G
SDP
Catalog No. 5000412268
Encompass_Preferred
Diisopropylammonium dichloroacetate is an organic compound used as a catalyst and ligand for various organic reactions, including olefin addition, carbonylation, and asymmetric catalytic reactions. It is also applied in medical fields such as anti-tumor therapy and serves as a biochemical reagent for life science research.
  • Catalyst and ligand for organic reactions
  • Promotes olefin addition, carbonylation reaction, and asymmetric catalytic reaction
  • Used in research for medical fields like anti-tumor therapy
  • Biochemical reagent for life science research
Catalog No. 50-004-12268 Supplier Medchemexpress LLC Supplier No. HYB2210500G
Diisopropylammonium dichloroacetate is an organic compound used as a catalyst and ligand for various organic reactions, including olefin addition, carbonylation, and asymmetric catalytic reactions. It is also applied in medical fields such as anti-tumor therapy and serves as a biochemical reagent for life science research.
  • Catalyst and ligand for organic reactions
  • Promotes olefin addition, carbonylation reaction, and asymmetric catalytic reaction
  • Used in research for medical fields like anti-tumor therapy
  • Biochemical reagent for life science research