Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylbenzene, 2:1 Mixture of m- and p-isomer 98%, Thermo Scientific™

Catalog No. AC407810010
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC407810010 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407810010 Supplier Thermo Scientific Chemicals Supplier No. 407810010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25321-09-9
Density 0.85g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C12H18
Refractive Index 1.4880 to 1.4910 (20°C, 589nm)
Linear Formula [(CH3)2CH)2C6H4
Quantity 1kg
MDL Number MFCD00054263
Synonym 1,2-diisopropylbenzene, diisopropylbenzene, o-diisopropylbenzene, benzene, 1,2-bis 1-methylethyl, benzene, o-diisopropyl, benzene, bis 1-methylethyl, unii-6e5460r9hg, diisopropylbenzene mixture of isomers, 1,2-bis 1-methylethyl benzene, diisopropyl benzene
InChI Key OKIRBHVFJGXOIS-UHFFFAOYSA-N
SMILES CC(C)C1=CC=CC=C1C(C)C
IUPAC Name 1,2-bis(propan-2-yl)benzene
Molecular Weight (g/mol) 162.28
PubChem CID 11345
Formula Weight 162.27
Percent Purity meta + para-diisopropylbenzene (GC):%gt; 97.5%
Chemical Name or Material Diisopropylbenzene, 98%
Show More Show Less
EINECSNumber 246-835-6
RTECSNumber CZ6330000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.