Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylphosphine, 10 wt.% solution in hexanes

Catalog No. p-3212630
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC383400025 2.5 g Glass bottle
AC383400100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC383400025 Supplier Thermo Scientific Chemicals Supplier No. 383400025
Only null left
Add to Cart

CAS: 20491-53-6 | C6H15P | 118.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropylphosphine
CAS 20491-53-6, 110-54-3
Color Brown-Yellow to Yellow
Density 0.6700g/mL
Flash Point −22°C
Packaging Glass bottle
Molecular Formula C6H15P
Linear Formula [(CH3)2CH]2PH
Quantity 2.5 g
MDL Number MFCD00233814
Specific Gravity 0.67
Synonym diisopropylphosphine, di-isopropylphosphine, di-i-propylphosphine, diisopropylphosphane, phosphine, diisopropyl, phosphine, bis 1-methylethyl, ipr2ph, di-iso-propylphosphine, di-iso-propyl phosphine, di propan-2-yl phosphane
Solubility Information Solubility in water: insoluble
InChI Key WDIIYWASEVHBBT-UHFFFAOYSA-N
SMILES CC(C)PC(C)C
IUPAC Name di(propan-2-yl)phosphane
Molecular Weight (g/mol) 118.16
PubChem CID 536496
Formula Weight 118.16
Percent Purity 9 to 11% (Total base)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.