Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisopropylphosphoramidous Dichloride

Catalog No. 3072545G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-725-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 30-725-45G Supplier Sigma Aldrich Supplier No. 3072545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the synthesis of peptides and oligonucleotide analogues.

Specifications

CAS 40216-83-9
Molecular Formula C6H11NO3 -+ HCl
Refractive Index n20/D 1.485
Linear Formula C6H11NO3 -+ HCl
MDL Number MFCD00080855
Quantity 5 g
Synonym (Diisopropylamino)dichlorophosphine; Bis(isopropyl)aminodichlorophosphine; Dichloro(diisopropylamido)phosphorus; Dichloro(diisopropylamino)phosphine
Molecular Weight (g/mol) 181.62 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.