Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisopropylzinc Solution

Catalog No. 5681125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
4 x 25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-811-25ML 5 mL
56-811-2 4 x 25 mL
56-811-225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 56-811-25ML Supplier Sigma Aldrich Supplier No. 5681125ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisopropylzinc is a general reagent used in the organic asymmetric synthesis such as the asymmetric synthesis of [6]?helicene, [5]-helicene and, enantiomerically enriched 5-pyrimidyl alkanol.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 625-81-0
Density 0.895 g/mL at 25°C
Molecular Formula C6H14Zn
Linear Formula Zn(CH(CH3)2)2
MDL Number MFCD01862461
Quantity 5 mL
Molecular Weight (g/mol) 151.57 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.