Learn More
Medchemexpress LLC Diketone-PEG4-biotin | 2353409-85-3 | 95.0% | C33H50N4O9S | 10MG
SDP
Catalog No. 5000260176
Encompass_Preferred
Diketone-PEG4-Biotin is a PEG-based PROTAC linker bearing a biotin affinity tag for use in chemical biology and targeted protein degradation research. It provides a flexible, water-soluble spacer to enable conjugation and affinity capture in the synthesis of bifunctional molecules and assay workflows.
  • Peg-based linker with biotin for affinity capture.
  • Compatible with PROTAC synthesis and bifunctional conjugates.
  • Provides a flexible spacer to reduce steric hindrance.
  • Facilitates pull-down assays and target engagement studies.
  • Supplied with specified purity for research applications.
  • Available in small research pack sizes for method development.
Catalog No. 50-002-60176 Supplier Medchemexpress LLC Supplier No. HY14044810MG
Diketone-PEG4-Biotin is a PEG-based PROTAC linker bearing a biotin affinity tag for use in chemical biology and targeted protein degradation research. It provides a flexible, water-soluble spacer to enable conjugation and affinity capture in the synthesis of bifunctional molecules and assay workflows.
  • Peg-based linker with biotin for affinity capture.
  • Compatible with PROTAC synthesis and bifunctional conjugates.
  • Provides a flexible spacer to reduce steric hindrance.
  • Facilitates pull-down assays and target engagement studies.
  • Supplied with specified purity for research applications.
  • Available in small research pack sizes for method development.