Learn More
Medchemexpress LLC Diketone-PEG4-PFP ester | 2353409-84-2 | ≥97.0% | 647.58 | 100 MG
SDP
Catalog No. 5000349448
Encompass_Preferred
Diketone-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49448 Supplier Medchemexpress LLC Supplier No. HY140006100MG
Diketone-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins