Learn More
Medchemexpress LLC Diketone-PEG4-PFP ester | 2353409-84-2 | ≥97.0% | 647.58 | 25 MG
SDP
Catalog No. 5000350905
Encompass_Preferred
Diketone-PEG4-PFP ester is a PROTAC linker, specifically a PEG-based PROTAC linker, designed for use in the synthesis of PROTACs. PROTACs themselves are compounds that contain two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • Diketone-PEG4-PFP ester is a PEG-based PROTAC linker.
  • It is used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective degradation of target proteins.
Catalog No. 50-003-50905 Supplier Medchemexpress LLC Supplier No. 25MGHY140006
Diketone-PEG4-PFP ester is a PROTAC linker, specifically a PEG-based PROTAC linker, designed for use in the synthesis of PROTACs. PROTACs themselves are compounds that contain two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • Diketone-PEG4-PFP ester is a PEG-based PROTAC linker.
  • It is used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective degradation of target proteins.