Learn More
Fine Science Tools DILATING FORCEPS,0.1MM,12CM
SDP
Catalog No. NC1006036
Encompass
Product: 12cm/0.1mm Tip Diameter/ Tip Shape: Angled 10 deg./ Tip Diameter: 0.1mm/ Alloy: Stainless Steel/ Length: 12cm/ Unit: Each
Catalog No. NC1006036 Supplier Fine Science Tools Supplier No. 1813112
Product: 12cm/0.1mm Tip Diameter/ Tip Shape: Angled 10 deg./ Tip Diameter: 0.1mm/ Alloy: Stainless Steel/ Length: 12cm/ Unit: Each