Learn More
Echelon Biosciences Research Labs DILAUROYL-SN-GLYCEROL 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L0012250MG
Encompass
Dilauroyl-sn-glycerol is a diacylglycerol (DAG) is involved in phospholipid metabolism and is a substrate of diacylglycerol kinase. 1,2-Diacylglycerols are formed in cells through the hydrolysis of phosphatidylinositol 4,5-bisphosphate by Phospholipase C. DAGs activate Protein Kinase C (PKC), which is a key regulator of signal transduction, cellular regulation, and tumor promotion. This item is non returnable
Catalog No. 50-720-460