Learn More
Medchemexpress LLC HY-100957 10mg Medchemexpress, Dilazep (dihydrochloride) CAS:20153-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10095710MG
Encompass_Preferred
Medchemexpress, HY-100957 10mg Dilazep (diHCl) CAS:20153-98-4 Dilazep dihydrochloride is an inhibitor of adenosine uptake. Dilazep dihydrochloride has cerebral and coronary vasodilating action through enhancement of effect of adenosine. Dilazep dihydrochloride also inhibits the ischemic damage, platelet aggregation, and membrane transport of nucleosides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7795