Learn More
Apexbio Technology LLC Dilazep dihydrochloride 20153-98-4 50mg
SDP
Catalog No. 5000092213
Encompass_Preferred
Dilazep dihydrochloride is a small-molecule inhibitor targeting adenosine reuptake It is designed to block the cellular uptake of adenosine thereby increasing extracellular adenosine concentrations and regulating adenosine receptor-mediated pathways Dilazep dihydrochloride exerts its biological activity primarily through inhibition of adenosine reuptake resulting in vasodilation and suppression of platelet aggregation Based on these pharmacological properties Dilazep dihydrochloride holds research potential in studies of coronary blood flow regulation platelet aggregation mechanisms cardiomyocyte metabolism and protective cellular responses during ischemia-reperfusion injury
Catalog No. 50-000-92213 Supplier Apexbio Technology LLC Supplier No. B631650
Dilazep dihydrochloride is a small-molecule inhibitor targeting adenosine reuptake It is designed to block the cellular uptake of adenosine thereby increasing extracellular adenosine concentrations and regulating adenosine receptor-mediated pathways Dilazep dihydrochloride exerts its biological activity primarily through inhibition of adenosine reuptake resulting in vasodilation and suppression of platelet aggregation Based on these pharmacological properties Dilazep dihydrochloride holds research potential in studies of coronary blood flow regulation platelet aggregation mechanisms cardiomyocyte metabolism and protective cellular responses during ischemia-reperfusion injury