Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilazep dihydrochloride, Thermo Scientific™

Catalog No. AAJ62459MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62459MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62459MF Supplier Thermo Scientific Chemicals Supplier No. J62459MF

Inhibits adenosine uptake and platelet aggregation; coronary vasodilator

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20153-98-4
Molecular Formula C31H46Cl2N2O10
Molecular Weight (g/mol) 677.61
MDL Number MFCD00133267
InChI Key VILIWRRWAWKXRW-UHFFFAOYSA-N
Synonym dilazep dihydrochloride, cormelian, asta c 4898, prestwick_175, unii-lv48lw10eo, lv48lw10eo, 3-4-3-3,4,5-trimethoxybenzoyl oxypropyl-1,4-diazepan-1-yl propyl 3,4,5-trimethoxybenzoate, 3-4-3-3,4,5-trimethoxybenzoyl oxypropyl-1,4-diazepane-1,4-diium-1-yl propyl 3,4,5-trimethoxybenzoate dichloride, dilazep dihydrochloride, powder, 1,4-bis 3-3,4,5-trimethoxybenzoyloxy propyl homopiperazine dihydrochloride
PubChem CID 6419915
IUPAC Name dihydrogen 3-{4-[3-(3,4,5-trimethoxybenzoyloxy)propyl]-1,4-diazepan-1-yl}propyl 3,4,5-trimethoxybenzoate dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC(OC)=C1OC)C(=O)OCCCN1CCCN(CCCOC(=O)C2=CC(OC)=C(OC)C(OC)=C2)CC1

Specifications

Color White
Quantity 50 mg
Merck Index 143200
Formula Weight 677.62
Physical Form Powder
Chemical Name or Material Dilazep dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 243-548-8
RTECSNumber DI0250000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.