Learn More
Enzo Life Sciences DilC12(3) perchlorate (ultra pure) (100mg). CAS: 75664-01-6
SDP
Catalog No. 502012625
Encompass_Preferred
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. The fluorescence of these environmentsensitive dyes is greatly enhanced when incorporated into membranes or bound to lipophilic biomolecules such as proteins although they are weakly fluorescent in water. Once applied to cells, these dyes diffuse laterally within the cellular plasma membranes, resulting in even staining of the entire cell at their optimal concentrations. DiIC12(3) is a lipophilic neuronal tracer that is commonly used for labeling neuronal projections as well as lipid bilayers in other cell types, particularly endothelial cells. Wavelength Maxima: Excitation 549nm, Emission 565nm. Alternative name: 1,1'-Didodecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2625 Supplier Enzo Life Sciences Supplier No. ENZ52206
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. The fluorescence of these environmentsensitive dyes is greatly enhanced when incorporated into membranes or bound to lipophilic biomolecules such as proteins although they are weakly fluorescent in water. Once applied to cells, these dyes diffuse laterally within the cellular plasma membranes, resulting in even staining of the entire cell at their optimal concentrations. DiIC12(3) is a lipophilic neuronal tracer that is commonly used for labeling neuronal projections as well as lipid bilayers in other cell types, particularly endothelial cells. Wavelength Maxima: Excitation 549nm, Emission 565nm. Alternative name: 1,1'-Didodecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.