Learn More
Biotium Dilinoleyl DiI
Catalog No. 501964728
Encompass_Preferred
Dilinoleyl DiI (1,1'-dilinoleyl-3,3,3',3'-tetramethylindocarbocynanine perchlorate), also known as FAST DiI, has identical absorption and emission spectra as DiI . Due to its unsaturated hydrocarbon chains, dilinoleyl DiI has a much faster lateral diffusion rate in cell membranes than DiI . This property makes dilinoleyl DiI particularly useful for tracing neurons in tissues. Properties: Ex/Em (MeOH) = 549/565 nm; e = 148,000; Dark red oily paste soluble in ethanol, DMF or DMSO; Store at -20°C and protect from light; C59H89ClN2O4; MW: 925.82 Fast DiI is a trademark of Molecular Probes.
Catalog No. 50-196-4728 Supplier Biotium Supplier No. 60034
Dilinoleyl DiI (1,1'-dilinoleyl-3,3,3',3'-tetramethylindocarbocynanine perchlorate), also known as FAST DiI, has identical absorption and emission spectra as DiI . Due to its unsaturated hydrocarbon chains, dilinoleyl DiI has a much faster lateral diffusion rate in cell membranes than DiI . This property makes dilinoleyl DiI particularly useful for tracing neurons in tissues. Properties: Ex/Em (MeOH) = 549/565 nm; e = 148,000; Dark red oily paste soluble in ethanol, DMF or DMSO; Store at -20°C and protect from light; C59H89ClN2O4; MW: 925.82 Fast DiI is a trademark of Molecular Probes.